Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Qorpak™ Clear Polystyrene Snap Cap Vials

Offers rigidity and glass-like clarity

Specifications

Closure Type Screw Cap
Color Clear
Material Polystyrene
For Use With (Application) Storing bench top accessories and field sampling
Quantity 144 Case
View More Specs

Products 3
Catalog Number Mfr. No. Volume (Metric) Diameter (Metric) Outer Height (Metric) Includes Price Quantity & Availability  
Catalog Number Mfr. No. Volume (Metric) Diameter (Metric) Outer Height (Metric) Includes Price Quantity & Availability  
02-991-774
View Documents
Qorpak™
PLC03732
29 mL 27 mm 29mm white LDPE unlined cap This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
02-992-605
View Documents
Qorpak™
PLC03734
33 mL 35 mm 36mm white LDPE unlined cap This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
02-991-501
View Documents
Qorpak™
PLC03731
25 mL 50 mm 25mm white LDPE unlined cap This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description

Description

  • Caps snap on/off easily
  • For storing bench top accessories and field sampling

Specifications

Specifications

Screw Cap
Polystyrene
144 Case
Clear
Storing bench top accessories and field sampling
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit