Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Qorpak™ Clear Square Tablet Bottles with Green Thermoset F217 and PTFE Cap

Ideal for mixing, storing and sampling of contents. Qorpak™ Clear Square Tablet Bottles with Green Thermoset F217 and PTFE Cap are space-saving and easy to fill.

Manufacturer:  Qorpak™ GLC01302

 View more versions of this product

Catalog No. 02-992-898


  Check Availability
Add to cart

Description

Description

  • Provides maximum content visibility
  • Feature wide mouth
Specifications

Specifications

2 oz., 60mL
2 oz.
F217 and PTFE
1.65 in.
Glass
Green
42mm
Screw
60mL
Thermoset
Clear
3.35 in.
144/Cs.
33-400
85mm
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit