Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Qorpak™ Clear Standard Wide Mouth Bottles, Ultra Clean

Feature a proprietary cleaning process that removes up to 99.5% of particles that are 5 microns and larger. Qorpak™ Clear Standard Wide Mouth Bottles, Ultra Clean are used for general lab use or fluid sampling.

Manufacturer:  Qorpak™ GLC01772

 View more versions of this product

Catalog No. 02-992-989


  Check Availability
Add to cart

Description

Description

  • Available with caps made of Thermoset F217 and PTFE
Specifications

Specifications

8 oz., 240mL
8 oz.
F217 and PTFE
2.44 in.
Glass
Green
62mm
Screw
240mL
Thermoset
Clear
4.29 in.
24/Cs.
58-400
109mm
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit