Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Qorpak™ Clear Standard Wide Mouth Bottles, Ultra Clean Available on GSA/VA Contract for Federal Government customers only.

Feature a proprietary cleaning process that removes up to 99.5% of particles that are 5 microns and larger. Qorpak™ Clear Standard Wide Mouth Bottles, Ultra Clean are used for general lab use or fluid sampling.

Manufacturer:  Qorpak™ GLC01884

 View more versions of this product

Catalog No. 02-992-987


  Check Availability
Add to cart

Description

Description

  • Available with caps made of Thermoset F217 and PTFE

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Specifications

128 oz., 3840mL
128 oz.
F217 and PTFE
6.18 in.
Glass
Green
157mm
Screw
3840mL
Thermoset
Clear
10.08 in.
4/Cs.
89-400
256mm
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit