Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Qorpak™ Clear Standard Wide Mouth Bottles without Cap

Ideal general purpose bottle. Qorpak™ Clear Standard Wide Mouth Bottles without Cap provides maximum visibility and integrity of the sample contents.

Manufacturer:  Qorpak™ GLA00873

 View more versions of this product

Catalog No. 02-991-906


  Check Availability
Add to cart

Description

Description

  • Features wide opening
  • Applications include pharmaceutical, chemical, environmental, and food packaging and storing
Specifications

Specifications

8 oz., 240mL
8 oz.
2.44 in.
Glass
24/Cs.
62mm
Screw
240mL
Clear
4.29 in.
Qorpak Carton
58-400
109mm
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit