Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Qorpak™ Clear Straight Sided Round Bottles with Black Phenolic Solid PE Cap

Designed without shoulders for maximum storage capacity

Manufacturer:  Qorpak™ GLC01619

 View more versions of this product

Catalog No. 02-992-716


  Check Availability
Add to cart

Description

Description

  • Multipurpose use
  • Provides content visibility

Specifications

Specifications

2 oz., 60mL
2 oz.
Polyethylene
2.17 in.
Glass
Black
55mm
Screw
60mL
Phenolic
Clear
1.93 in.
144/Cs.
53-400
49mm
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit