Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Qorpak™ Clear Straight-Sided Round Bottles, Cleaned for Semi-Volatiles with White Caps

Cleaned for semi-volatiles including pesticides, PCB's and metals

Manufacturer:  Qorpak™ GLC01654

 View more versions of this product

Catalog No. 02-992-043


  Check Availability
Add to cart

Description

Description

  • Provide maximum storage capacity and content visibility
  • Designed without shoulders
  • Made of clear glass

Certifications

Meets U.S. EPA Specifications

Specifications

Specifications

8 oz., 240mL
8 oz.
PTFE
2.2 in.
Glass
58-400
137mm
240mL
Polypropylene
Clear
5.39 in.
White
56mm
Screw
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit