Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Qorpak™ Clear Straight Sided Round Bottles with Phenolic Pulp/Aluminum Foil Lined Cap

Provides maximum storage capacity and content visibility. Qorpak™ Clear Straight Sided Round Bottles with Phenolic Pulp/Aluminum Foil Lined Cap are made of clear glass.

Manufacturer:  Qorpak™ GLC04875

 View more versions of this product

Catalog No. 02-992-262


  Check Availability
Add to cart

Description

Description

  • Designed without shoulders
Specifications

Specifications

6 oz., 180mL
6 oz.
Pulp/Aluminum Foil
2.56 in.
Glass
63-400
79mm
180mL
Phenolic
Clear
3.11 in.
Black
65mm
Screw
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit