Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Qorpak™ Clear Straight Sided Round Bottles with Thermoset F217 and PTFE Lined Caps

Offers maximum content visibility

Manufacturer:  Qorpak™ GLC01631

 View more versions of this product

Catalog No. 02-993-221


Add to cart

Description

Description

  • Without shoulders for maximum storage capacity
  • Green cap easily identifies PTFE liner system
  • Usage: analytical lab samples, high purity chemicals, strong acids solvents, environmental samples, pharmaceuticals, and diagnostic reagents

Specifications

Specifications

4 oz., 120mL
4 oz.
F217 and PTFE
2.36 in.
Glass
Green
60mm
Screw
120mL
Thermoset
Clear
2.68 in.
24/Cs.
58-400
68mm
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit