Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.

Qorpak™ Clear Straight-Sided Round Bottles — With Phenolic Solid PE Caps

Odor-free and tasteless with clean appearance

Manufacturer:  Qorpak™ GLC01727

 View more versions of this product

Catalog No. 02-992-923



Description

Description

  • Provides maximum storage capacity and content visibility
  • Designed without shoulders
  • Made of clear glass
  • Separate washing not necessary
  • Vacuum and ionized

Specifications

Specifications

32 oz., 960mL
32 oz.
Polyethylene
3.74 in.
Glass
12/Cs.
89-400
170mm
960mL
Phenolic
Clear
6.69 in.
Conventional Pack
Black
95mm
Screw
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit