Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Qorpak™ Clear Straight Sided Ultra Clean Round Bottles

For containers free from particulate contaminates

Manufacturer:  Qorpak™ GLC07454

 View more versions of this product

Catalog No. 02-992-657


Add to cart

Description

Description

  • Provide maximum storage capacity and content visibility
  • Designed without shoulders
  • Made of clear glass
  • Cleaned using three-step process to remove traces of bottle release reagents

Specifications

Specifications

16 oz., 480mL
16 oz.
F217 and PTFE
3.58 in.
Glass
89-400
96mm
480mL
Thermoset
Clear
3.78 in.
Green
91mm
Screw
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit