Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.
Qorpak™ Clear Wide Mouth French Square Bottles, Ultra Clean Available on GSA/VA Contract for Federal Government customers only.
Designed for those needing containers cleaned for general lab use or fluid sampling. Qorpak™ Clear Wide Mouth French Square Bottles, Ultra Clean feature a proprietary cleaning process that removes up to 99.5% of particles that are 5 microns and larger.
Manufacturer: Qorpak™ GLC01262
 View more versions of this product
Catalog No. 02-992-431
  N/A
Add to Cart
Related Products
Product Content Feedback