Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Qorpak™ Clear Wide Mouth French Square Bottles, Ultra Clean Available on GSA/VA Contract for Federal Government customers only.

Designed for those needing containers cleaned for general lab use or fluid sampling. Qorpak™ Clear Wide Mouth French Square Bottles, Ultra Clean feature a proprietary cleaning process that removes up to 99.5% of particles that are 5 microns and larger.

Manufacturer:  Qorpak™ GLC01313

 View more versions of this product

Catalog No. 02-992-446


  Check Availability
Add to cart

Description

Description

  • Made of clear glass
  • Provide maximum content visibility
  • Designed to save shelf and storage space
  • Attached caps reduce carton dust
  • No traces of bottle release agents
  • Ideal for mixing, storing and sampling

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Specifications

4 oz., 120mL
4 oz.
F217 and PTFE
1.77 in.
Glass
Green
45mm
Screw
120mL
Thermoset
Clear
4.41 in.
24/Cs.
33-400
112mm
Documents
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit