Learn More
Qorpak™ Clear French Square Bottles without Cap
Ideal for mixing, storing and sampling of contents. Qorpak™ Clear French Square Bottles without Cap is space saving and easy to fill.
Manufacturer:  Qorpak™ GLA00822
 View more versions of this product
Catalog No. 02-993-222
  Check Availability
Add to cart
Description
Description
  • Features wide mouth
  • Clear glass provides maximum content visibility and minimal chemical inference with bottle’s contents
  • Designed to save shelf and storage space
Specifications
Specifications
0.5 oz., 15mL
0.5 oz.
0.94 in.
Glass
20-400
62mm
15mL
Clear
2.44 in.
48/Cs.
24mm
Screw