Qorpak™ CurTec™ Lids for White Packo Containers Available on GSA/VA Contract for Federal Government customers only.

For use with Packo containers. Qorpak™ CurTec™ Lids provides high level of product protection and added value through quality, functionality, ease of use, and design.

Manufacturer: Qorpak™ 248748

 View more versions of this product

Catalog No. 02-994-110

  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Specifications

    Product Type CurTec Lid
    Color Red
    No. per Pack 30 Case
    Packaging Convenience
    For Use With 300 to 1300mL white pack containers
    Material Polypropylene

    • Made of polypropylene
    • Availabe with and without gasket

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit