Learn More
Qorpak™ CurTec™ Wide Neck Drums Available on GSA/VA Contract for Federal Government customers only.
Provides high level of product protection and added value through quality, functionality, ease of use, and design. Qorpack™ CurTec™ Wide Neck Drums are used for shipping and stocking valuable fine chemicals.
Manufacturer:  Qorpak™ 248733
 View more versions of this product
Catalog No. 02-994-117
  Check Availability
Add to cart
Description
Description
  • Features red polypropylene unlined lid
  • Wide neck for easy filling of contents
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
HACCP, ISO
10.7 in.
Ideal for pharmaceuticals, fine chemicals, food ingredients and hazardous waste
417mm
14.5kg
20L
Cap
274mm
16.4 in.
32 lb.
676 oz.
White