Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Qorpak™ Glass Bottle Beakers with Green Thermoset F217 and PTFE Caps
Click to view available options
Capacity:
1 oz., 30mL
16 oz., 480 mL
2 oz., 60 mL
4 oz., 120 mL
8 oz., 240 mL
Height (Metric):
68 mm
83 mm
102 mm
127 mm
145 mm
Height (English):
2.68 in.
3.27 in.
4.02 in.
5 in.
5.71 in.
Quantity:
144/Cs.
24/Cs.
288/Cs.
432/Cs.
48/Cs.
96/Cs.
Description
- Designed with slight shoulder for easy pouring and handling
- Molded graduations in mL and ounces
- Made of clear glass
- For use with biological and pathological specimens
- For storage of industrial laboratory chemicals and reagents
Specifications
Closure Liner | F217, PTFE |
Closure Material | Thermoset |
Closure Size | 48-400 mm |
Closure Type | Screw Cap |
Diameter (Metric) | 51 mm |
Height (Metric) | 102 mm |
Height (English) | 4.02 in. |
Capacity | 4 oz., 120 mL |
Capacity (Metric) | 120 mL |
Capacity (English) | 4 oz. |
Show More |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction