Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Qorpak™ Graduated Glass Bottle Beakers with Black Phenolic Pulp/Vinyl Caps
Click to view available options
Capacity:
1 oz., 30mL
16 oz., 480 mL
2 oz., 60 mL
4 oz., 120 mL
8 oz., 240 mL
Packaging:
Bulk Pack
Conventional Pack
Quantity:
144/Cs.
24/Cs.
288/Cs.
432/Cs.
48/Cs.
96/Cs.
Description
- Designed with slight shoulder for easy pouring and handling
- Molded graduations in mL and ounces
- Made of clear glass
- For use with biological and pathological specimens
- For storage of industrial laboratory chemicals and reagents
Specifications
Diameter (Metric) | 42 mm |
Closure Liner | Pulp/Vinyl |
Closure Material | Phenolic |
Closure Size | 38-400 mm |
Closure Type | Screw Cap |
Height (English) | 3.27 in. |
Height (Metric) | 83 mm |
Capacity | 2 oz., 60 mL |
Capacity (Metric) | 60 mL |
Capacity (English) | 2 oz. |
Show More |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction