Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Qorpak™ Graduated Medium Round Bottles, Vacuum and Ionized with Black Phenolic Pulp/Vinyl Caps

Odor-free and tasteless with no need to clean before initial use

Manufacturer:  Qorpak™ GLC01554

 View more versions of this product

Catalog No. 02-992-168


  Check Availability
Add to cart

Description

Description

  • Made of clear (flint) glass
  • Graduated in both milliliters and ounces
  • Designed with slight shoulder for easy pouring and handling
  • Wide mouth facilitates easy mixing, handling and transfer of contents
  • Removal of contaminants such as loose dirt, carton lint, aerosols and fine glass particles

Specifications

Specifications

8 oz., 240mL
8 oz.
Phenolic
Clear
5 in.
Conventional Pack
Black
62mm
Screw
240mL
Vacuum and ionized
F217 and PTFE
2.44 in.
Glass
24/Cs.
58-400
127mm
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit