Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Qorpak Graduated Medium Round Bottles With Metal Plastisol Cap
Mix, measure and store in the same container
Manufacturer: Qorpak GLC01567
Description
- Designed with slight shoulder for easy pouring and handling
- Molded graduations in mL and ounces
- Made of clear glass
- For use with biological and pathological specimens
- For storage of industrial laboratory chemicals and reagents
Specifications
16 oz., 480mL | |
16 oz. | |
Clear | |
5.71 in. | |
24/Cs. | |
70-400 | |
145mm |
480mL | |
Plastisol | |
2.99 in. | |
Glass | |
White | |
76mm | |
Screw |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title