Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Qorpak™ Graduated Medium Round Bottles — With Metal Plastisol Cap

Mix, measure and store in the same container

Manufacturer:  Qorpak™ GLC06375

 View more versions of this product

Catalog No. 02-992-176


Add to cart

Description

Description

  • Designed with slight shoulder for easy pouring and handling
  • Molded graduations in mL and ounces
  • Made of clear glass
  • For use with biological and pathological specimens
  • For storage of industrial laboratory chemicals and reagents
Specifications

Specifications

4 oz., 120mL
4 oz.
Clear
4.02 in.
24/Cs.
48-400
102mm
120mL
Plastisol
2.01 in.
Glass
White
51mm
Screw
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit