Qorpak™ Green Thermoset F217 and PTFE Lined Caps Available on GSA/VA Contract for Federal Government customers only.

Offer wide range of chemical compatibility and high temperature tolerance

Manufacturer: Qorpak™ CAP00547

 View more versions of this product

Catalog No. 02-991-146

  Check Availability
Add to Cart

    Specifications

    Closure Liner F217 and PTFE
    Closure Type Ribbed
    Color Green
    For Use With Narrow mouth bottle
    Neck Finish 15-425
    No. per Case 11000 Cs.
    Packaging Bulk

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit