Learn More
Qorpak™ Green Thermoset F217 and PTFE Lined Caps Available on GSA/VA Contract for Federal Government customers only.
Offer wide range of chemical compatibility and high temperature tolerance
Manufacturer:  Qorpak™ CAP00549
 View more versions of this product
Catalog No. 02-991-208
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
F217 and PTFE
6500 Cs.
Wide mouth bottle
Smooth
Green
Bulk
18-400