Learn More
Qorpak™ Green Thermoset F217 and PTFE Lined Caps
Offer wide range of chemical compatibility and high temperature tolerance
Manufacturer:  Qorpak™ CAP00550
 View more versions of this product
Catalog No. 02-993-429
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
  • Resist attack from chemicals at room temperature
  • Can be used when odor and impact resilient tight seal is required
  • With low moisture transmission rate
  • Available in ribbed and smooth styles
  • Available in bagged or bulk packing
Specifications
Specifications
F217 and PTFE
500 Cs.
Bagged
18-400
Green
100/Pk.
Wide mouth bottle
Ribbed