Learn More
Qorpak™ Green Thermoset F217 and PTFE Lined Caps Available on GSA/VA Contract for Federal Government customers only.
Offer wide range of chemical compatibility and high temperature tolerance
Manufacturer:  Qorpak™ CAP00558
 View more versions of this product
Catalog No. 36-101-0434
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
F217 and PTFE
500 Cs.
Bagged
28-400
Green
100/Pk.
Wide mouth bottle
Smooth