Learn More
Qorpak HDPE Pail Lids
For pails of 1, 2, 3.5, 5 and 6 gal. volumes
Manufacturer: Qorpak PLA03294
Description
- Available in plain, Senior Rieke spout and EZ Tear styles
Rieke spout and Senior Rieke spout lids:
- Facilitate dispensing of liquid contents
- Feature pull tabs that secure contents during shipping and transport
Specifications
White | |
24.91 cm | |
Plain Lid | |
0.69 in. | |
200/Cs. |
9.81 in. | |
HDPE | |
2 gal. pails | |
1.75 cm |