Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Qorpak™ Kaptclean™ Amber Borosilicate Vials, Cleaned for Volatiles: With White Polypropylene PTFE/Silicone Septa Caps

Offer protection from harmful light and UV rays for light sensitive products

Manufacturer:  Qorpak™ GLC07868

 View more versions of this product

Catalog No. 02-992-566

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »


Description

Description

  • Made of Type I borosilicate glass, providing resistance to heat shock and chemical leaching
  • For use in volatile organic analysis

Certifications

Meet U.S. EPA Specifications and Guidelines

Specifications

Specifications

Polypropylene
1.09 in.
Cleaned for volatiles
140mm
Screw
15 dram
27.5 mm
60 mL
Screw cap
Use in volatile organic analysis
5.51 in.
72/Cs.
Cleaned Sample Vial
Amber
Borosilicate Glass
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit