Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Qorpak™ Kaptclean™ Amber Borosilicate Vials, Cleaned for Volatiles: With White Polypropylene PTFE/Silicone Septa Caps Available on GSA/VA Contract for Federal Government customers only.

Offer protection from harmful light and UV rays for light sensitive products

$231.00 - $485.00

Specifications

Closure Type Screw Cap
Diameter (Metric) Outer 27.7 mm
Color Amber
Material Borosilicate Glass
For Use With (Application) Use in volatile organic analysis
View More Specs

Products 5
Catalog Number Mfr. No. Volume (Metric) Type Height (Metric) Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Volume (Metric) Type Height (Metric) Quantity Price Quantity & Availability  
02-992-220 Available on GSA/VA Contract for Federal Government customers only.
View Documents
Qorpak™
GLC01057
20 mL 57 mm 80/Cs.
Case of 80 for $238.00
Add to cart
 
02-992-300 Available on GSA/VA Contract for Federal Government customers only.
View Documents
Qorpak™
GLC01071
40 mL 95 mm 144/Cs.
Case of 144 for $485.00
Add to cart
 
02-992-316 Available on GSA/VA Contract for Federal Government customers only.
View Documents
Qorpak™
GLC01076
40 mL 95 mm 144/Cs.
Case of 144 for $468.00
Add to cart
 
02-992-489 Available on GSA/VA Contract for Federal Government customers only.
View Documents
Qorpak™
GLC01059
20 mL 57 mm 144/Cs.
Case of 80 for $231.00
Add to cart
 
02-992-022
View Documents
Qorpak™
GLC07869
60 mL 140 mm 72/Cs.
Case of 72 for $452.00
Add to cart
 
Description

Description

  • Made of Type I borosilicate glass, providing resistance to heat shock and chemical leaching
  • For use in volatile organic analysis

Certifications

Meet U.S. EPA Specifications and Guidelines

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Specifications

Screw Cap
Amber
Use in volatile organic analysis
Screw
27.7 mm
Borosilicate Glass
Polypropylene
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit