Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Qorpak™ Kaptclean™ Clear Borosilicate Vials, Ready to Clean Available on GSA/VA Contract for Federal Government customers only.

Highly resistant to heat shock and chemical leaching

$282.00 - $338.00

Specifications

Closure Type Screw Cap
Color Clear
Material Borosilicate Glass
For Use With (Application) Highly resistant to heat and chemicals
Closure Material Polypropylene
View More Specs

Products 4
Catalog Number Mfr. No. Volume (Metric) Height (English) Quantity Volume (English) Price Quantity & Availability  
Catalog Number Mfr. No. Volume (Metric) Height (English) Quantity Volume (English) Price Quantity & Availability  
02-991-474
View Documents
Qorpak™
GLC05184
40 mL 3.18 in. 144/Cs. 10 dr.
Case of 144 for $338.00
Add to cart
 
02-991-826
View Documents
Qorpak™
GLC05080
40 mL 3.74 in. 144/Cs. 10 dr.
Case of 144 for $282.00
Add to cart
 
02-992-934 Available on GSA/VA Contract for Federal Government customers only.
View Documents
Qorpak™
GLC04835
20 mL 2.26 in. 144/Cs. 5 dr.
Case of 144 for $323.00
Add to cart
 
02-992-866
View Documents
Qorpak™
GLC07880
60 mL 5.51 in. 72/cs. 15 dr.
Case of 72 for $325.00
Add to cart
 
Description

Description

  • With high visibility
  • Available with polypropylene caps and friction fit 0.015 in. (0.38mm) inert PTFE disc
  • Both cap and liner can be washed, heated or autoclaved
  • Available in volumes ranging from 5 to 15 dr. (20 to 60mL)

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Specifications

Screw Cap
Borosilicate Glass
Polypropylene
Clear
Highly resistant to heat and chemicals
Screw
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit