Learn More
Qorpak™ LDPE Cubitainers with White PP SturdeeSeal™ PE Foam Lined Caps Available on GSA/VA Contract for Federal Government customers only.
Non-glug construction facilitates flow of contents om continuous uninterrupted streams
Manufacturer:  Qorpak™ PLA03242
 View more versions of this product
Catalog No. 02-991-315
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
  • Designed to collapse as product is dispensed
  • Reversible well design permits opening to be raised or depressed for pouring and storage
  • Can withstand rigors of hazardous chemical packaging
  • 128 and 640 oz. (3.7 and 19L) containers include heavy-duty corrugated outer carton
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
128 oz., 3.7L
For storage and transport of liquid
128 oz.
3.78L
No
Natural
Cube
Cubitainer
LDPE
10/Cs.