Learn More
Qorpak™ LDPE Cubitainers with White PP SturdeeSeal™ PE Foam Lined Caps Available on GSA/VA Contract for Federal Government customers only.
Non-glug construction facilitates flow of contents om continuous uninterrupted streams
Manufacturer:  Qorpak™ PLC03457
 View more versions of this product
Catalog No. 02-991-153
  Check Availability
Add to cart
Description
Description
  • Designed to collapse as product is dispensed
  • Reversible well design permits opening to be raised or depressed for pouring and storage
  • Can withstand rigors of hazardous chemical packaging
  • 128 and 640 oz. (3.7 and 19L) containers include heavy-duty corrugated outer carton
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
32 oz., 1L
Natural
Cube
Cubitainer
LDPE
144/Cs.
Polypropylene
For storage and transport of liquid
32 oz.
0.94L
No