Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Qorpak™ Metal F-Style Oblong Cans - With Alpha Solvseal Lined Cap and Inner Seal Available on GSA/VA Contract for Federal Government customers only.

Compact structure enables easy fit on shelves

Manufacturer:  Qorpak™ MET03048

 View more versions of this product

Catalog No. 02-991-240

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

  • Air tight seal provided during original filling and can readily be achieved when resealing after use
  • Shock and temperature resistant
  • May be sterilized by heat
  • Available with or without handle
  • Also available without Alpha Solvseal Lined Cap and Inner Seal

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Specifications

1.74 in.
For cooking oils, paint thinners, turpentine, lighter fluids, insecticides or general storage applications
907mm
24.01 in.
6Case
Oblong
403mm
3785mL
Metal
4.44cm
35.7 in.
Alpha solvseal lined cap and innerseal, handle
610mm
F Style Can
15.86 in.
128 oz.
Silver
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit