Learn More
Qorpak™ Natural HDPE Square Bottles Available on GSA/VA Contract for Federal Government customers only.
Suggested for chemical, pharmaceutical, cosmetic and food applications. Qorpak? HDPE Square Bottles feature a space-saving design.
Manufacturer:  Qorpak™ 241565
 View more versions of this product
Catalog No. 02-994-002
  Check Availability
Add to cart
Specifications
Description
  • Wide neck for easy filling
  • HDPE construction offers chemical compatibility
Description & Specifications
Specifications
250mL
Natural
HDPE
280/Cs.