Qorpak™ Natural HDPE Square Bottles Available on GSA/VA Contract for Federal Government customers only.
Suggested for chemical, pharmaceutical, cosmetic and food applications. Qorpak™ HDPE Square Bottles feature a space-saving design.
Manufacturer: Qorpak™ 241566
 View more versions of this product
Catalog No. 02-994-003
  Check Availability
Add to Cart
    Specifications
    Volume (Metric) 500mL
    Color Natural
    Material HDPE
    Quantity 150/Cs.
    • Wide neck for easy filling
    • HDPE construction offers chemical compatibility
  • Description & Specifications
Related Products
Product Content Feedback