Learn More
Qorpak™ Natural HDPE Wide Mouth Oblong Bottles — Without Caps Available on GSA/VA Contract for Federal Government customers only.
Offer a space saving design for the workbench. Qorpak™ Natural HDPE Wide Mouth Oblong Bottles — without Cap are ideal for high-speed filling, demanding pharmaceutical specifications and the rough handling of interstate shipping.
Manufacturer:  Qorpak™ PLA03252
 View more versions of this product
Catalog No. 02-991-642
  Check Availability
Add to cart
Description
Description
  • Feature a wide mouth opening, ideal for sampling solids and powders
  • HDPE material provides moisture barrier and are impact resistant
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
2 oz., 60mL
2 oz.
2.83 in.
Bulk Pack
33-400
43mm
33-400
60mL
Natural
HDPE
950/Cs.
1.69 in.
72mm
Screw