Learn More
Qorpak™ Natural HDPE Wide Mouth Round Bottles with White Polypropylene PE Foam Cap Available on GSA/VA Contract for Federal Government customers only.
Ideal for general lab use with solids and powders. Qorpak™ Natural HDPE Wide Mouth Round Bottles with White Polypropylene PE Foam Cap provide better moisture barrier and are impact resistant.
Manufacturer:  Qorpak™ PLC03548
 View more versions of this product
Catalog No. 02-992-940
  Check Availability
Add to cart
Description
Description
  • For high-speed filling, demanding pharmaceutical specifications and rough handling of interstate shipping
  • Standard style available in sizes from 32 oz. to 128 oz.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
3840mL (128 oz.)
PE Foam
HDPE
89-400
Polypropylene
Natural
White