Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Qorpak™ Natural HDPE Wide Mouth Round Bottles with Black Phenolic Solid PE Caps, Vacuum and Ionized Available on GSA/VA Contract for Federal Government customers only.

Individually cleaned of contaminants and sealed

Manufacturer:  Qorpak™ PLC03617

 View more versions of this product

Catalog No. 02-993-258

  Check Availability
Add to cart

Specifications Description & Specifications

Specifications

68 oz., 2000mL
68 oz.
Polyethylene
8.5 in.
Black
3.1 in.
217mm
Screw
2000mL
Phenolic
Natural
HDPE
89-400
124mm
89-400
Promotions

Promotions

  • Simport Scientific , Saint Gobain Performance Plastics , Qorpak , Bel-Art Products , Fisherbrand
    Get the essentials you need at a price you'll love.

    12-31-2017

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit