Learn More
Qorpak™ Natural HDPE Wide Mouth Round Bottles with PP SturdeeSeal™ PE Foam Caps, Cleaned for Volatiles Available on GSA/VA Contract for Federal Government customers only.
For general lab use with solids and powders
Manufacturer:  Qorpak™ PLC03611
 View more versions of this product
Catalog No. 02-991-393
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
32.12 oz., 950mL
32.12 oz.
SturdeeSeal™ Polyethylene Foam
HDPE
White
95mm
53-400
950mL
Polypropylene
Natural
12/Cs.
53-400
180mm
Screw