Learn More
Qorpak™ Natural HDPE Wide Mouth Round Bottles with PP SturdeeSeal™ PE Foam Caps, Cleaned for Volatiles Available on GSA/VA Contract for Federal Government customers only.
For general lab use with solids and powders
Manufacturer:  Qorpak™ PLC03643
 View more versions of this product
Catalog No. 02-991-399
  Check Availability
Add to cart
Description
Description
  • Provide moisture barrier
  • Translucent
  • Impact resistant
  • For high-speed filling, demanding pharmaceutical specifications and rough handling of interstate shipping
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Certifications
Meets U.S. EPA Specifications and Guidelines for Contaminant-Free Sample Containers
Specifications
Specifications
4 oz., 120mL
4 oz.
SturdeeSeal™ Polyethylene Foam
HDPE
White
47mm
38-400
120mL
Polypropylene
Natural
48/Cs.
38-400
95mm
Screw