Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Qorpak™ Natural HDPE Wide Mouth Round Bottles — With Black Phenolic Pulp/Vinyl Caps Available on GSA/VA Contract for Federal Government customers only.

Provide moisture barrier

Manufacturer:  Qorpak™ PLC03605

 View more versions of this product

Catalog No. 02-991-571


  Check Availability
Add to cart

Specifications Description & Specifications

Specifications

32.12 oz., 950mL
32.12 oz.
Pulp/Vinyl
7.1 in.
Conventional Pack
Black
3.7 in.
180mm
Screw
950mL
Phenolic
Natural
HDPE
12/Cs.
53-400
95mm
53-400
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit