Learn More
Qorpak™ Natural HDPE Wide Mouth Round Bottles without Caps Available on GSA/VA Contract for Federal Government customers only.
Ideal for general use with solids and powders
Manufacturer:  Qorpak™ PLA03315
 View more versions of this product
Catalog No. 02-991-088
  Check Availability
Add to cart
Description
Description
  • Provides moisture barrier
  • Impact resistant
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
2 oz., 60mL
2 oz.
2.9 in.
850/Cs.
1.6 in.
75mm
Screw
60mL
Natural
HDPE
33-400
40mm
33-400