Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Qorpak™ Phenolic and Polypropylene Hole Caps
Click to view available options
Neck Finish:
13-425
15-425
18-400
20-400
22-400
24-400
24-410
Material:
Phenolic Resin
Polypropylene
No. per Case:
100 Cs.
10500 Cs.
17000 Cs.
3700 Cs.
4000 Cs.
500 Cs.
5300 Cs.
576 Cs.
5800 Cs.
7500 Cs.
Packaging:
Bagged
Bulk
Description
Phenolic Hole Caps
- Pliable
- Chemically compatible
- Most temperature tolerant among plastic caps
- Provide consistent adherence to close dimensional tolerances
- Ideal for pharmaceutical applications
- Septa sold separately
- Pliable
- Septa sold separately
Specifications
For Use With (Equipment) | Wide mouth bottle |
Product Type | Hole Cap |
Color | Black |
Material | Phenolic Resin |
No. per Case | 5800 Cs. |
Packaging | Bulk |
Neck Finish | 22-400 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction