Qorpak™ PTFE Sealing Septa and Discs Available on GSA/VA Contract for Federal Government customers only.

Easy-to-clean, remove and replace

Manufacturer: Qorpak™ CAP00576

 View more versions of this product

Catalog No. 02-993-124

  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Specifications

    Thickness (Metric) 0.25mm
    Thickness (English) 0.01 in.
    Color Natural
    For Use With Bottle closure
    Material Polytetrafluoroethylene
    Neck Finish 89-400
    No. per Case 1000 Cs.
    Packaging Bulk

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit