Learn More
Qorpak PTFE Sealing Septa and Discs
Easy-to-clean, remove and replace
Manufacturer: Qorpak CAP00530
Description
Septa
- PTFE/Silicone
- Resists coring
- Provides great resealability
- Good for multiple injections
- PTFE resistant until punctured, once punctured the septa is silicone compatible
- Converts ordinary caps into noncontaminating closures
Specifications
Natural | |
500 Cs. | |
0.25mm | |
Bottle closure |
PTFE | |
Bagged | |
0.01 in. | |
45-400 |