Learn More
Qorpak™ PTFE Sealing Septa and Discs Available on GSA/VA Contract for Federal Government customers only.
Easy-to-clean, remove and replace
Manufacturer:  Qorpak™ CAP00526
 View more versions of this product
Catalog No. 02-993-165
  Check Availability
Add to cart
Description
Description
Septa
  • PTFE/Silicone
  • Resists coring
  • Provides great resealability
  • Good for multiple injections
  • PTFE resistant until punctured, once punctured the septa is silicone compatible
Discs
  • Converts ordinary caps into noncontaminating closures
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Natural
500 Cs.
0.25mm
Bottle closure
PTFE
Bagged
0.01 in.
38-400