Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.
Qorpak™ Sterile Polypropylene Hinged Vials Available on GSA/VA Contract for Federal Government customers only.
Do not require second means of sterilization
Manufacturer:  Qorpak™ PLC10057
 View more versions of this product
Catalog No. 02-991-606
Description
Description
  • Leak-proof and airtight
  • Manufactured in Class 10,00 and Class 100,000 clean rooms
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Leakproof
50mm
250 Case
4 oz.
63mm
Sterile
Yes
Lock seal tab
Hinged Vial
Natural
Polypropylene
120mL