Learn More
Qorpak™ Sterile Polypropylene Hinged Vials Available on GSA/VA Contract for Federal Government customers only.
Do not require second means of sterilization
Manufacturer:  Qorpak™ PLC10054
 View more versions of this product
Catalog No. 02-991-202
  Check Availability
Add to cart
Description
Description
  • Leak-proof and airtight
  • Manufactured in Class 10,00 and Class 100,000 clean rooms
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Sterile
90mL
33mm
3 oz.
Hinged Vial
Break off arrow and lock seal tab
Tamper evident, leakproof
3 oz., 90mL
Natural
Polypropylene
93mm
400 Case