Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Qorpak™ Trigger Sprayer Pumps
Click to view available options
No. per Case:
144 Cs.
375 Cs.
Packaging:
Bagged
Bulk
Description
- Made of white polypropylene
- Available with PE Foam liner or industrial sprayers with strainer,stainless-steel balls and springs
Pump with F217 Foam PE liner
- For general use trigger spray bottles
- For industrial chemical trigger spray bottles
- Chemical resistant
Specifications
Closure Liner | F217 Foam Polyethylene |
For Use With (Equipment) | Trigger spray bottle |
Color | White |
Material | Polypropylene |
No. per Case | 375 Cs. |
Packaging | Bulk |
Neck Finish | 28-400 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction