Learn More
Qorpak™ Unlined Round Cans with Triple Tite Lid Available on GSA/VA Contract for Federal Government customers only.
Permeation protection against light, oxygen, other gases and essential oils
Manufacturer:  Qorpak™ MET03098
 View more versions of this product
Catalog No. 02-991-248
  Check Availability
Add to cart
Description
Description
  • Features resealable multi-friction plug lid for more than one point of contact between lid and can
  • Provides an airtight seal when opened by prying and sealed by pressing
  • May be sterilized by heat
  • Used for paints, varnishes, pastes and adhesives
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
6.625 in.
7.5 in.
Triple tite lid
Open Top Can
3785mL
Metal
For paints, varnishes, pastes and adhesives, protection against light, oxygen, other gases and essential oil
708mm
46 Case
128 oz.
Silver