Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Qorpak™ Unlined Round Cans with Triple Tite Lid
Permeation protection against light, oxygen, other gases and essential oils
Supplier: Qorpak™ MET03101
Description
- Features resealable multi-friction plug lid for more than one point of contact between lid and can
- Provides an airtight seal when opened by prying and sealed by pressing
- May be sterilized by heat
- Used for paints, varnishes, pastes and adhesives
Specifications
Open Top Can | |
3785 mL | |
For paints, varnishes, pastes and adhesives, protection against light, oxygen, other gases and essential oil | |
Triple Tite Lid, Ears, Bails and Securing Clips | |
27.87 in. | |
24.01 in. |
128 oz. | |
Silver | |
Metal | |
46 Cs. | |
708 mm | |
610 mm |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction